2-(Cyclopropylmethoxy)-4-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

015882-500MG
101803-916EA 278.46 USD
101803-916
2-(Cyclopropylmethoxy)-4-fluorobenzaldehyde ≥95%
2-(Cyclopropylmethoxy)-4-fluorobenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₁FO₂
MDL Number: MFCD06246757
CAS Number: 883525-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR