2-Ethoxy-4-fluorobenzaldehyde ≥95%
Supplier: Matrix Scientific
015880-500MG
101803-914EA
278.46
USD
101803-914
2-Ethoxy-4-fluorobenzaldehyde ≥95%
2-Ethoxy-4-fluorobenzaldehyde
Hazard: Irritant
Formula:
C₉H₉FO₂ |
MDL Number:
MFCD06247345 CAS Number: 883537-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...