4-(Cyclopropylmethoxy)-1-naphthaldehyde

Supplier: Matrix Scientific

015810-500MG
101803-908EA 278.46 USD
101803-908
4-(Cyclopropylmethoxy)-1-naphthaldehyde
4-(Cyclopropylmethoxy)-1-naphthaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MDL Number: MFCD06246797
CAS Number: 883528-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR