5-Benzyl-2-methoxybenzaldehyde
Supplier: Matrix Scientific
015783-500MG
101803-896EA
278.46
USD
101803-896
5-Benzyl-2-methoxybenzaldehyde
5-Benzyl-2-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₅H₁₄O₂ Boiling Pt: 229…230 °C (20 Torr) Melting Pt: 55 °C |
MDL Number:
MFCD06245849 CAS Number: 722493-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...