4,5-Dimethoxy-2-methylbenzaldehyde

Supplier: Matrix Scientific

015775-500MG
101803-894EA 278.46 USD
101803-894
4,5-Dimethoxy-2-methylbenzaldehyde
4,5-Dimethoxy-2-methylbenzaldehyde

Hazard: Irritant/Stored under Argon

Formula: C₁₀H₁₂O₃
Boiling Pt: 110…111 °C (0.2 Torr)
Melting Pt: 76 °C
MDL Number: MFCD00094433
CAS Number: 7721-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR