5-(tert-Butyl)-2-methoxybenzenecarbaldehyde

Supplier: Matrix Scientific

Synonyms: 5-(tert-Butyl)-2-methoxybenzaldehyde

015751-500MG
101803-888EA 278.46 USD
101803-888
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 128 °C (3.5 Torr)
Melting Pt: 32 °C
MDL Number: MFCD06246086
CAS Number: 85943-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR