4-Ethoxy[1,1'-biphenyl]-3-carbaldehyde
Supplier: Matrix Scientific
015744-500MG
101803-886EA
278.46
USD
101803-886
4-Ethoxy[1,1'-biphenyl]-3-carbaldehyde
4-Ethoxy[1,1'-biphenyl]-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₅H₁₄O₂ |
MDL Number:
MFCD06247318 CAS Number: 883535-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...