3-(Cyclopropylmethoxy)benzaldehyde

Supplier: Matrix Scientific

015678-500MG
101803-874EA 278.46 USD
101803-874
3-(Cyclopropylmethoxy)benzaldehyde
3-(Cyclopropylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD06246720
CAS Number: 58986-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR