3-(tert-Butyl)-5-methyl-4-propoxybenzaldehyde

Supplier: Matrix Scientific

015657-500MG
101803-866EA 278.46 USD
101803-866
3-(tert-Butyl)-5-methyl-4-propoxybenzaldehyde
3-(tert-Butyl)-5-methyl-4-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₂₂O₂
MDL Number: MFCD06246091
CAS Number: 883515-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR