3-Chloro-4-methoxybenzaldehyde
Supplier: Matrix Scientific
015587-500MG
101803-856EA
278.46
USD
101803-856
3-Chloro-4-methoxybenzaldehyde
3-Chloro-4-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₈H₇ClO₂ MW: 170.6 g/mol Boiling Pt: 128 °C (5 Torr) Melting Pt: 53 °C |
MDL Number:
MFCD00016601 CAS Number: 4903-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...