4-Ethoxy-5,6,7,8-tetrahydro-1-naphthalenecarbaldehyde

Supplier: Matrix Scientific

015576-500MG
101803-852EA 335.38 USD
101803-852
4-Ethoxy-5,6,7,8-tetrahydro-1-naphthalenecarbaldehyde
4-Ethoxy-5,6,7,8-tetrahydro-1-naphthalenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
MDL Number: MFCD06247414
CAS Number: 883540-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR