4-Methoxy-5,6,7,8-tetrahydro-1-naphthalenecarbaldehyde

Supplier: Matrix Scientific

015575-500MG
101803-850EA 321.54 USD
101803-850
4-Methoxy-5,6,7,8-tetrahydro-1-naphthalenecarbaldehyde
4-Methoxy-5,6,7,8-tetrahydro-1-naphthalenecarbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD06248104
CAS Number: 883531-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR