2-(Methoxymethyl)piperidine

Supplier: Matrix Scientific

015565-500MG
101803-842EA 430.77 USD
101803-842
2-(Methoxymethyl)piperidine
2-(Methoxymethyl)piperidine

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD02663567
CAS Number: 104678-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR