1-Ethoxy-2-propanamine hydrochloride

Supplier: Matrix Scientific

015532-500MG
101803-816EA 278.46 USD
101803-816
1-Ethoxy-2-propanamine hydrochloride
1-Ethoxy-2-propanamine hydrochloride

Hazard: Irritant

Formula: C₅H₁₄ClNO
MDL Number: MFCD09879167
CAS Number: 1185304-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR