2-(3-Methoxypropoxy)-N-methyl-1-propanamine
Supplier: Matrix Scientific
015244-500MG
101803-804EA
278.46
USD
101803-804
2-(3-Methoxypropoxy)-N-methyl-1-propanamine
2-(3-Methoxypropoxy)-N-methyl-1-propanamine
Hazard: Irritant
Formula:
C₈H₁₉NO₂ |
MDL Number:
MFCD06248093 CAS Number: 883531-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...