2-(3-Methoxypropoxy)-N-methyl-1-ethanamine
Supplier: Matrix Scientific
015243-500MG
101803-802EA
278.46
USD
101803-802
2-(3-Methoxypropoxy)-N-methyl-1-ethanamine
2-(3-Methoxypropoxy)-N-methyl-1-ethanamine
Hazard: Irritant
Formula:
C₇H₁₇NO₂ |
MDL Number:
MFCD06248085 CAS Number: 883545-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...