4-(Tetrahydro-2-furanylmethoxy)piperidine hydrochloride
Supplier: Matrix Scientific
015017-500MG
101803-800EA
278.46
USD
101803-800
4-(Tetrahydro-2-furanylmethoxy)piperidine hydrochloride
4-(Tetrahydro-2-furanylmethoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClNO₂ |
MDL Number:
MFCD09475408 CAS Number: 1174044-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...