2-(Tetrahydro-2-furanylmethoxy)-1-propanamine

Supplier: Matrix Scientific

015002-500MG
101803-796EA 278.46 USD
101803-796
2-(Tetrahydro-2-furanylmethoxy)-1-propanamine
2-(Tetrahydro-2-furanylmethoxy)-1-propanamine

Hazard: Irritant

Formula: C₈H₁₇NO₂
MDL Number: MFCD06248806
CAS Number: 883537-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR