2-(Tetrahydro-2-furanylmethoxy)-1-ethanamine

Supplier: Matrix Scientific

015001-500MG
101803-794EA 278.46 USD
101803-794
2-(Tetrahydro-2-furanylmethoxy)-1-ethanamine
2-(Tetrahydro-2-furanylmethoxy)-1-ethanamine

Hazard: Irritant

Formula: C₇H₁₅NO₂
Boiling Pt: 162…164 °C (740 Torr)
MDL Number: MFCD06248788
CAS Number: 34694-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR