2-Isobutoxy-1-ethanamine hydrochloride

Supplier: Matrix Scientific

014951-500MG
101803-790EA 278.46 USD
101803-790
2-Isobutoxy-1-ethanamine hydrochloride
2-Isobutoxy-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₆H₁₆ClNO
MDL Number: MFCD09037389
CAS Number: 1181458-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR