3-[(Neopentyloxy)methyl]piperidine

Supplier: Matrix Scientific

014914-500MG
101803-786EA 278.46 USD
101803-786
3-[(Neopentyloxy)methyl]piperidine
3-[(Neopentyloxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₁H₂₃NO
MDL Number: MFCD06248580
CAS Number: 883544-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR