4-(3-Phenylpropoxy)piperidine hydrochloride

Supplier: Matrix Scientific

014692-500MG
101803-784EA 278.46 USD
101803-784
4-(3-Phenylpropoxy)piperidine hydrochloride
4-(3-Phenylpropoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD09442326
CAS Number: 745066-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR