4-[(3-Phenylpropoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
014691-500MG
101803-782EA
278.46
USD
101803-782
4-[(3-Phenylpropoxy)methyl]piperidine hydrochloride
4-[(3-Phenylpropoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD09064912 CAS Number: 1051919-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...