3-[(3-Phenylpropoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

014689-500MG
101803-780EA 278.46 USD
101803-780
3-[(3-Phenylpropoxy)methyl]piperidine hydrochloride
3-[(3-Phenylpropoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD08447192
CAS Number: 1050509-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR