N,N-Dimethyl-3-(3-piperidinylmethoxy)-1-propanamine hydrochloride

Supplier: Matrix Scientific

014614-500MG
101803-776EA 278.46 USD
101803-776
N,N-Dimethyl-3-(3-piperidinylmethoxy)-1-propanamine hydrochloride
N,N-Dimethyl-3-(3-piperidinylmethoxy)-1-propanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅ClN₂O
MDL Number: MFCD09475407
CAS Number: 1093653-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR