2-Propoxy-1-butanamine
Supplier: Matrix Scientific
014579-500MG
101803-770EA
278.46
USD
101803-770
2-Propoxy-1-butanamine
2-Propoxy-1-butanamine
Hazard: Irritant
Formula:
C₇H₁₇NO |
MDL Number:
MFCD06248731 CAS Number: 883532-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...