2-(3,3-Dimethylbutoxy)-1-ethanamine

Supplier: Matrix Scientific

014551-500MG
101803-768EA 278.46 USD
101803-768
2-(3,3-Dimethylbutoxy)-1-ethanamine
2-(3,3-Dimethylbutoxy)-1-ethanamine

Hazard: Irritant

Formula: C₈H₁₉NO
MDL Number: MFCD06247137
CAS Number: 883525-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR