4-[(Cyclopentylmethoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

014416-500MG
101803-746EA 278.46 USD
101803-746
4-[(Cyclopentylmethoxy)methyl]piperidine hydrochloride
4-[(Cyclopentylmethoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO
MDL Number: MFCD09879164
CAS Number: 1185299-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR