2-(Cyclopropylmethoxy)-N-methyl-1-ethanamine

Supplier: Matrix Scientific

014318-500MG
101803-742EA 278.46 USD
101803-742
2-(Cyclopropylmethoxy)-N-methyl-1-ethanamine
2-(Cyclopropylmethoxy)-N-methyl-1-ethanamine

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD06246780
CAS Number: 883526-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR