2-(Cyclopropylmethoxy)-1-ethanamine

Supplier: Matrix Scientific

014301-500MG
101803-736EA 438.46 USD
101803-736
2-(Cyclopropylmethoxy)-1-ethanamine
2-(Cyclopropylmethoxy)-1-ethanamine

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD06246745
CAS Number: 883524-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR