4-(3,4-Difluorophenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
014042-500MG
101803-730EA
278.46
USD
101803-730
4-(3,4-Difluorophenoxy)piperidine hydrochloride
4-(3,4-Difluorophenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄ClF₂NO |
MDL Number:
MFCD08544114 CAS Number: 204013-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...