4-[(1-Naphthyloxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

013991-500MG
101803-720EA 278.46 USD
101803-720
4-[(1-Naphthyloxy)methyl]piperidine hydrochloride
4-[(1-Naphthyloxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClNO
MDL Number: MFCD08445798
CAS Number: 1185294-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR