N-Methyl-2-(4-methylphenoxy)-1-ethanamine

Supplier: Matrix Scientific

013893-500MG
101803-700EA 278.46 USD
101803-700
N-Methyl-2-(4-methylphenoxy)-1-ethanamine
N-Methyl-2-(4-methylphenoxy)-1-ethanamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD06248189
CAS Number: 625437-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR