3-[(4-Methylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

013889-500MG
101803-692EA 208.15 USD
101803-692
3-[(4-Methylphenoxy)methyl]piperidine
3-[(4-Methylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD06248328
CAS Number: 883547-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR