2-([1,1'-Biphenyl]-4-yloxy)ethylamine hydrochloride

Supplier: Matrix Scientific

013801-500MG
101803-688EA 278.46 USD
101803-688
2-([1,1'-Biphenyl]-4-yloxy)ethylamine hydrochloride
2-([1,1'-Biphenyl]-4-yloxy)ethylamine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₆ClNO
MDL Number: MFCD08445694
CAS Number: 1051368-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR