N-[2-(3,4-Dimethylphenoxy)ethyl]-N-methylamine

Supplier: Matrix Scientific

013743-500MG
101803-674EA 278.46 USD
101803-674
N-[2-(3,4-Dimethylphenoxy)ethyl]-N-methylamine
N-[2-(3,4-Dimethylphenoxy)ethyl]-N-methylamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD06247221
CAS Number: 883529-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR