2-(3,4-Dimethylphenoxy)-1-propanamine hydrochloride

Supplier: Matrix Scientific

013727-500MG
101803-672EA 278.46 USD
101803-672
2-(3,4-Dimethylphenoxy)-1-propanamine hydrochloride
2-(3,4-Dimethylphenoxy)-1-propanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClNO
MDL Number: MFCD08720552
CAS Number: 1050508-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR