N-Methyl-2-(3-methylphenoxy)-1-ethanamine hydrochloride

Supplier: Matrix Scientific

013719-500MG
101803-670EA 278.46 USD
101803-670
N-Methyl-2-(3-methylphenoxy)-1-ethanamine hydrochloride
N-Methyl-2-(3-methylphenoxy)-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO
MDL Number: MFCD08727962
CAS Number: 1050509-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR