3-[(3-Methylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

013715-500MG
101803-664EA 278.46 USD
101803-664
3-[(3-Methylphenoxy)methyl]piperidine hydrochloride
3-[(3-Methylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD08447190
CAS Number: 28569-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR