2-(3-Fluorophenoxy)-N-methyl-1-ethanamine hydrochloride
Supplier: Matrix Scientific
013669-500MG
101803-656EA
278.46
USD
101803-656
2-(3-Fluorophenoxy)-N-methyl-1-ethanamine hydrochloride
2-(3-Fluorophenoxy)-N-methyl-1-ethanamine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClFNO |
MDL Number:
MFCD08721749 CAS Number: 1050508-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...