3-[(3-Fluorophenoxy)methyl]piperidine
Supplier: Matrix Scientific
013665-500MG
101803-650EA
278.46
USD
101803-650
3-[(3-Fluorophenoxy)methyl]piperidine
3-[(3-Fluorophenoxy)methyl]piperidine
Hazard: Irritant
Formula:
C₁₂H₁₆FNO |
MDL Number:
MFCD06247687 CAS Number: 405090-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...