4-(2-Benzylphenoxy)piperidine

Supplier: Matrix Scientific

013643-500MG
101803-648EA 278.46 USD
101803-648
4-(2-Benzylphenoxy)piperidine
4-(2-Benzylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₈H₂₁NO
MDL Number: MFCD06245924
CAS Number: 883511-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR