4-[(2-Benzylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

013642-500MG
101803-646EA 278.46 USD
101803-646
4-[(2-Benzylphenoxy)methyl]piperidine hydrochloride
4-[(2-Benzylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD09859622
CAS Number: 1093653-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR