2-Chloro-5-methylphenyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

013467-500MG
101803-626EA 278.46 USD
101803-626
2-Chloro-5-methylphenyl-4-piperidinylmethyl ether hydrochloride
2-Chloro-5-methylphenyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD08445713
CAS Number: 1185294-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR