2-(2-Chloro-4-fluorophenoxy)ethylamine hydrochloride

Supplier: Matrix Scientific

013427-500MG
101803-620EA 278.46 USD
101803-620
2-(2-Chloro-4-fluorophenoxy)ethylamine hydrochloride
2-(2-Chloro-4-fluorophenoxy)ethylamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₀Cl₂FNO
MDL Number: MFCD08728063
CAS Number: 1051919-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR