3-[(2-Chlorophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

013415-500MG
101803-614EA 278.46 USD
101803-614
3-[(2-Chlorophenoxy)methyl]piperidine hydrochloride
3-[(2-Chlorophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD08447187
CAS Number: 1018446-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR