2-(2-Fluorophenoxy)-1-ethanamine

Supplier: Matrix Scientific

013352-500MG
101803-604EA 278.46 USD
101803-604
2-(2-Fluorophenoxy)-1-ethanamine
2-(2-Fluorophenoxy)-1-ethanamine

Hazard: Irritant

Formula: C₈H₁₀FNO
MW: 155.17 g/mol
MDL Number: MFCD00235182
CAS Number: 120351-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR