4-(Perfluorocyclohexyl)butan-1-ol ≥97%

Supplier: Matrix Scientific

012870-1G
101803-548EA 127.69 USD
101803-548
4-(Perfluorocyclohexyl)butan-1-ol ≥97%
4-(Perfluorocyclohexyl)butan-1-ol

Hazard: Irritant

Formula: C₁₀H₉F₁₁O
MDL Number: MFCD06248652
CAS Number: 883546-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR