5,5,6,6,6-Pentafluorohexan-1-ol ≥97%

Supplier: Matrix Scientific

012864-1G
101803-538EA 89.23 USD
101803-538
5,5,6,6,6-Pentafluorohexan-1-ol ≥97%
5,5,6,6,6-Pentafluorohexan-1-ol

Hazard: Irritant

Formula: C₆H₉F₅O
MDL Number: MFCD06248629
CAS Number: 58556-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR