(2,3-Dibromopentafluoropropoxy)benzene ≥97%

Supplier: Matrix Scientific

012843-5G
101803-494EA 95.38 USD
101803-494
(2,3-Dibromopentafluoropropoxy)benzene ≥97%
(2,3-Dibromopentafluoropropoxy)benzene

Hazard: Irritant

Formula: C₉H₅Br₂F₅O
MDL Number: MFCD06246842
CAS Number: 883532-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR